Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131378.1 | NP_571453.1 | 4 - 5 | 671 | 90 | |
XM_021480025.1 | XP_021335700.1 | 4 - 5 | 692 | 90 | ||
GenBank mRNA | AF132085.2 | - | 4 - 5 | 671 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Danio rerio Probe spans exons |