Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313822.1 | NP_001300751.1 | 11 - 12 | 1774 | 97 | |
NM_131791.2 | NP_571866.2 | 10 - 11 | 1598 | 97 | ||
XM_005166892.4 | XP_005166949.1 | 11 - 12 | 1777 | 97 | ||
GenBank mRNA | AB042250.1 | - | 10 - 11 | 97 | ||
BC095305.1 | - | 11 - 12 | 1746 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |