Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153663.2 | NP_705949.2 | 12 - 13 | 1678 | 87 | |
XM_005163832.4 | XP_005163889.1 | 12 - 13 | 1692 | 87 | ||
GenBank mRNA | AF465772.1 | - | 12 - 13 | 1634 | 87 | |
BC048024.1 | - | 12 - 13 | 1633 | 87 | ||
BC165625.1 | - | 11 - 12 | 1478 | 87 | ||
CF999294.1 | - | 4 - 5 | 526 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |