Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v1e1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173254.1 | NP_775361.1 | 6 - 7 | 628 | 70 | |
GenBank mRNA | AF506222.1 | - | 6 - 7 | 628 | 70 | |
BC057254.1 | - | 6 - 7 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |