Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult1st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182941.1 | NP_891986.1 | 2 - 3 | 256 | 102 | |
GenBank mRNA | AY181064.1 | - | 2 - 3 | 256 | 102 | |
BC056729.1 | - | 2 - 3 | 170 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |