Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkcbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199641.1 | NP_955935.1 | 12 - 13 | 1737 | 134 | |
XM_005161943.4 | XP_005162000.1 | 13 - 14 | 1846 | 134 | ||
XM_005161944.4 | XP_005162001.1 | 13 - 14 | 2308 | 134 | ||
XM_005161945.4 | XP_005162002.1 | 13 - 14 | 2151 | 134 | ||
XM_005161946.4 | XP_005162003.1 | 13 - 14 | 1767 | 134 | ||
XM_009296662.3 | XP_009294937.1 | 13 - 14 | 1746 | 134 | ||
XM_009296664.3 | XP_009294939.1 | 13 - 14 | 1749 | 134 | ||
XM_009296665.3 | XP_009294940.1 | 13 - 14 | 1846 | 134 | ||
GenBank mRNA | BC058064.1 | - | 12 - 13 | 1737 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |