Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199662.1 | NP_955956.1 | 5 - 6 | 759 | 82 | |
GenBank mRNA | BC044445.1 | - | 5 - 6 | 759 | 82 | |
BC165559.1 | - | 4 - 5 | 679 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |