Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf3c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200262.1 | NP_956556.1 | 1 | 646 | 85 | |
GenBank mRNA | BC049013.1 | - | 1 | 646 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |