Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spag7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201125.1 | NP_957419.1 | 1 - 2 | 108 | 63 | |
GenBank mRNA | BC054697.1 | - | 1 - 2 | 108 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |