Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rapgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328346.1 | NP_001315275.1 | 5 - 6 | 824 | 67 | |
XM_021468780.1 | XP_021324455.1 | 5 - 6 | 947 | 67 | ||
XM_021468781.1 | XP_021324456.1 | 5 - 6 | 947 | 67 | ||
XM_021468782.1 | XP_021324457.1 | 5 - 6 | 945 | 67 | ||
XM_021468783.1 | XP_021324458.1 | 5 - 6 | 946 | 67 | ||
XM_021468784.1 | XP_021324459.1 | 5 - 6 | 944 | 67 | ||
GenBank mRNA | BC049305.1 | - | 5 - 6 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |