Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxsr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201175.3 | NP_957469.2 | 10 - 11 | 1102 | 59 | |
XM_021470117.1 | XP_021325792.1 | 10 - 11 | 1129 | 59 | ||
GenBank mRNA | BC044183.1 | - | 10 - 11 | 1088 | 59 | |
BC067372.1 | - | 10 - 11 | 1080 | 59 | ||
BC165393.1 | - | 10 - 11 | 986 | 59 | ||
EH577479.1 | - | 1 - 2 | 140 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |