Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201218.1 | NP_957512.1 | 2 - 3 | 158 | 104 | |
XM_009298160.3 | XP_009296435.1 | 3 - 4 | 1098 | 104 | ||
XM_009298162.3 | XP_009296437.1 | 3 - 4 | 1095 | 104 | ||
XM_021470468.1 | XP_021326143.1 | 3 - 4 | 1108 | 104 | ||
GenBank mRNA | AY423003.1 | - | 2 - 3 | 158 | 104 | |
BC114255.1 | - | 3 - 4 | 203 | 104 | ||
BC124462.1 | - | 3 - 4 | 449 | 104 | ||
BC134072.1 | - | 3 - 4 | 932 | 104 | ||
BC152529.1 | - | 3 - 4 | 932 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |