Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndel1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201344.1 | NP_958501.1 | 8 - 9 | 1046 | 109 | |
GenBank mRNA | BC055587.1 | - | 8 - 9 | 1046 | 109 | |
BC164385.1 | - | 7 - 8 | 973 | 109 | ||
EF133514.1 | - | 8 - 9 | 974 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |