Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205652.1 | 483 | 78 | |||
GenBank mRNA | BC056284.1 | - | 468 | 78 | ||
BC065662.1 | - | 483 | 78 |
Gene Symbol: wbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017354877.2 | XP_017210366.2 | 521 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio |