Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205687.2 | NP_991250.1 | 2 - 3 | 287 | 79 | |
XM_005169876.4 | XP_005169933.1 | 3 - 4 | 520 | 79 | ||
GenBank mRNA | AY424306.1 | - | 3 - 4 | 326 | 79 | |
AY522553.1 | - | 3 - 4 | 314 | 79 | ||
BC116611.1 | - | 2 - 3 | 287 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |