Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212604.3 | NP_997769.3 | 10 - 11 | 1221 | 109 | |
GenBank mRNA | BC056538.1 | - | 10 - 11 | 1107 | 109 | |
BC067624.1 | - | 10 - 11 | 1115 | 109 | ||
EH596437.1 | - | 2 - 3 | 701 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |