Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cstf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328559.1 | NP_001315488.1 | 4 - 5 | 711 | 152 | |
GenBank mRNA | BC054661.1 | - | 3 - 4 | 961 | 152 | |
BC066520.1 | - | 3 - 4 | 676 | 152 | ||
EH592404.1 | - | 4 - 5 | 718 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |