Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gng12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213312.2 | NP_998477.2 | 2 - 3 | 332 | 88 | |
XM_009297872.3 | XP_009296147.1 | 2 - 3 | 124 | 88 | ||
GenBank mRNA | BC068343.1 | - | 2 - 3 | 273 | 88 | |
EH439686.1 | - | 2 - 3 | 360 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |