Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214774.2 | NP_999939.1 | 2 - 3 | 440 | 61 | |
GenBank mRNA | BC055255.1 | - | 2 - 3 | 380 | 61 | |
BC097054.1 | - | 2 - 3 | 428 | 61 | ||
BC165114.1 | - | 2 - 3 | 312 | 61 | ||
BQ262702.1 | - | 2 - 3 | 348 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |