Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gria4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214806.1 | NP_999971.1 | 2 - 3 | 386 | 68 | |
XM_017359227.2 | XP_017214716.1 | 2 - 3 | 460 | 68 | ||
XM_017359228.2 | XP_017214717.1 | 2 - 3 | 465 | 68 | ||
XM_017359229.2 | XP_017214718.1 | 2 - 3 | 464 | 68 | ||
XM_021481376.1 | XP_021337051.1 | 2 - 3 | 473 | 68 | ||
GenBank mRNA | AF525747.1 | - | 2 - 3 | 386 | 68 | |
BC162576.1 | - | 2 - 3 | 314 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |