Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgef18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214818.1 | NP_999983.1 | 17 - 18 | 2118 | 125 | |
XM_005171284.4 | XP_005171341.1 | 16 - 17 | 2161 | 125 | ||
XM_009298637.3 | XP_009296912.1 | 18 - 19 | 2225 | 125 | ||
XM_021479667.1 | XP_021335342.1 | 17 - 18 | 2198 | 125 | ||
GenBank mRNA | BC070002.1 | - | 17 - 18 | 2118 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |