Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002060.1 | NP_001002060.1 | 2 - 3 | 342 | 90 | |
XM_009302036.3 | XP_009300311.1 | 2 - 3 | 416 | 90 | ||
GenBank mRNA | BC071346.1 | - | 2 - 3 | 342 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |