Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002060.1 | NP_001002060.1 | 6 - 7 | 748 | 79 | |
XM_009302036.3 | XP_009300311.1 | 6 - 7 | 855 | 79 | ||
GenBank mRNA | BC071346.1 | - | 6 - 7 | 748 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |