Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002091.1 | NP_001002091.1 | 3 - 4 | 181 | 76 | |
GenBank mRNA | BC071538.1 | - | 3 - 4 | 181 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |