Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002207.1 | NP_001002207.1 | 8 - 9 | 808 | 65 | |
GenBank mRNA | BC072706.1 | - | 8 - 9 | 808 | 65 | |
BC165883.1 | - | 8 - 9 | 837 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |