Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327829.1 | NP_001314758.1 | 3 - 4 | 333 | 103 | |
XM_005164872.4 | XP_005164929.1 | 2 - 3 | 370 | 103 | ||
XM_005164874.4 | XP_005164931.1 | 2 - 3 | 370 | 103 | ||
XM_009300373.3 | XP_009298648.1 | 4 - 5 | 590 | 103 | ||
XM_017354995.2 | XP_017210484.1 | 4 - 5 | 589 | 103 | ||
XM_017354996.2 | XP_017210485.1 | 4 - 5 | 585 | 103 | ||
GenBank mRNA | BC076355.1 | - | 2 - 3 | 341 | 103 | |
BC164744.1 | - | 1 - 2 | 197 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |