Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002667.1 | NP_001002667.1 | 3 - 4 | 649 | 90 | |
XM_021479882.1 | XP_021335557.1 | 4 - 5 | 773 | 90 | ||
GenBank mRNA | BC076521.1 | - | 3 - 4 | 649 | 90 | |
BC164094.1 | - | 3 - 4 | 321 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |