Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003477.1 | NP_001003477.1 | 1 - 2 | 370 | 107 | |
GenBank mRNA | BC078383.1 | - | 1 - 2 | 370 | 107 | |
BC164069.1 | - | 1 - 2 | 229 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |