Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ralba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003649.1 | NP_001003649.1 | 4 - 5 | 692 | 78 | |
XM_021478640.1 | XP_021334315.1 | 4 - 5 | 719 | 78 | ||
GenBank mRNA | BC078184.1 | - | 4 - 5 | 692 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |