Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003862.1 | NP_001003862.1 | 10 - 11 | 942 | 70 | |
GenBank mRNA | AY648755.1 | - | 10 - 11 | 942 | 70 | |
BC095222.1 | - | 10 - 11 | 945 | 70 | ||
BC165456.1 | - | 10 - 11 | 945 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |