Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003866.1 | NP_001003866.1 | 6 - 7 | 673 | 72 | |
GenBank mRNA | AY648780.1 | - | 6 - 7 | 673 | 72 | |
BC095553.1 | - | 6 - 7 | 671 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |