Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004110.1 | NP_001004110.1 | 22 - 23 | 2298 | 101 | |
XM_005158836.4 | XP_005158893.1 | 23 - 24 | 2552 | 101 | ||
XM_009293188.3 | XP_009291463.1 | 22 - 23 | 2543 | 101 | ||
XM_021467147.1 | XP_021322822.1 | 23 - 24 | 2556 | 101 | ||
XM_021467148.1 | XP_021322823.1 | 22 - 23 | 2546 | 101 | ||
GenBank mRNA | AY618701.1 | - | 22 - 23 | 2298 | 101 | |
AY691328.1 | - | 23 - 24 | 2451 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |