Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sephs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004295.2 | NP_001004295.2 | 4 - 5 | 787 | 59 | |
GenBank mRNA | AY221265.1 | - | 4 - 5 | 762 | 59 | |
BC081590.1 | - | 4 - 5 | 770 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |