Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020656.1 | NP_001018492.1 | 4 - 5 | 768 | 116 | |
XM_005168979.4 | XP_005169036.1 | 4 - 5 | 820 | 116 | ||
GenBank mRNA | BC095679.1 | - | 4 - 5 | 768 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |