Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: vtg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025189.2 | NP_001020360.2 | 15 - 16 | 2157 | 143 | |
XM_021469376.1 | XP_021325051.1 | 15 - 16 | 2151 | 143 | ||
GenBank mRNA | BC097081.1 | - | 15 - 16 | 2154 | 143 | |
EH532952.1 | - | 4 - 5 | 439 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |