Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025498.1 | NP_001020669.1 | 3 - 4 | 639 | 99 | |
XM_005160713.3 | XP_005160770.1 | 3 - 4 | 769 | 99 | ||
XM_005160714.3 | XP_005160771.1 | 3 - 4 | 763 | 99 | ||
XM_021468445.1 | XP_021324120.1 | 4 - 5 | 704 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |