Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025498.1 | NP_001020669.1 | 4 - 5 | 712 | 65 | |
XM_005160713.3 | XP_005160770.1 | 4 - 5 | 842 | 65 | ||
XM_005160714.3 | XP_005160771.1 | 4 - 5 | 836 | 65 | ||
XM_021468445.1 | XP_021324120.1 | 5 - 6 | 777 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |