Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030172.1 | NP_001025343.1 | 1 - 2 | 274 | 95 | |
GenBank mRNA | BC097141.1 | - | 1 - 2 | 274 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |