Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031838.1 | NP_001027008.1 | 10 - 11 | 1012 | 105 | |
XM_005161414.4 | XP_005161471.1 | 10 - 11 | 1005 | 105 | ||
XM_005161417.4 | XP_005161474.1 | 10 - 11 | 1005 | 105 | ||
GenBank mRNA | AY918066.1 | - | 10 - 11 | 1012 | 105 | |
BC057511.1 | - | 10 - 11 | 893 | 105 | ||
BC133108.1 | - | 10 - 11 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |