Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039637.1 | NP_001034726.1 | 15 - 16 | 1664 | 95 | |
XM_005166055.4 | XP_005166112.1 | 16 - 17 | 2050 | 95 | ||
XM_009302561.3 | XP_009300836.1 | 16 - 17 | 2052 | 95 | ||
XM_021476910.1 | XP_021332585.1 | 16 - 17 | 1749 | 95 | ||
XM_021476911.1 | XP_021332586.1 | 15 - 16 | 2010 | 95 | ||
XM_021476912.1 | XP_021332587.1 | 13 - 14 | 1842 | 95 | ||
XM_021476913.1 | XP_021332588.1 | 12 - 13 | 1405 | 95 | ||
XM_021476914.1 | XP_021332589.1 | 12 - 13 | 1502 | 95 | ||
XM_021476916.1 | XP_021332591.1 | 12 - 13 | 1639 | 95 | ||
XM_021476917.1 | XP_021332592.1 | 16 - 17 | 2048 | 95 | ||
XM_021476918.1 | XP_021332593.1 | 11 - 12 | 1378 | 95 | ||
GenBank mRNA | BC163445.1 | - | 12 - 13 | 1415 | 95 | |
DQ333303.2 | - | 15 - 16 | 1664 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |