Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn4aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039825.1 | NP_001034914.1 | 17 - 18 | 3039 | 105 | |
XM_017358479.2 | XP_017213968.1 | 18 - 19 | 3225 | 105 | ||
GenBank mRNA | DQ149506.1 | - | 17 - 18 | 3039 | 105 | |
DQ351532.1 | - | 6 - 7 | 1227 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |