Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039826.2 | NP_001034915.2 | 4 - 5 | 466 | 143 | |
GenBank mRNA | AB201384.1 | - | 4 - 5 | 441 | 143 | |
BC121730.1 | - | 4 - 5 | 466 | 143 | ||
BC163941.1 | - | 3 - 4 | 449 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |