Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039976.1 | NP_001035065.1 | 1 - 2 | 512 | 98 | |
GenBank mRNA | BC081679.1 | - | 1 - 2 | 518 | 98 | |
BC097105.1 | - | 1 - 2 | 512 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |