Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044311.2 | NP_001037776.2 | 2 - 3 | 320 | 62 | |
GenBank mRNA | BC090287.1 | - | 2 - 3 | 353 | 62 | |
BC117596.1 | - | 2 - 3 | 274 | 62 | ||
BC165594.1 | - | 1 - 2 | 268 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |