Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044311.2 | NP_001037776.2 | 7 - 8 | 1064 | 114 | |
GenBank mRNA | BC090287.1 | - | 7 - 8 | 1097 | 114 | |
BC117596.1 | - | 7 - 8 | 1018 | 114 | ||
BC165594.1 | - | 6 - 7 | 1012 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |