Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stim1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044799.1 | NP_001038264.1 | 9 - 10 | 1390 | 113 | |
XM_005173392.4 | XP_005173449.1 | 9 - 10 | 1440 | 113 | ||
XM_005173393.4 | XP_005173450.1 | 9 - 10 | 1440 | 113 | ||
XM_009291871.3 | XP_009290146.1 | 9 - 10 | 1440 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |