Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045207.1 | NP_001038672.1 | 24 - 25 | 3036 | 69 | |
XM_005158734.4 | XP_005158791.1 | 25 - 26 | 3066 | 69 | ||
XM_021467217.1 | XP_021322892.1 | 26 - 27 | 3093 | 69 | ||
XM_021467218.1 | XP_021322893.1 | 25 - 26 | 3084 | 69 | ||
XM_021467219.1 | XP_021322894.1 | 26 - 27 | 3093 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |