Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045207.1 | NP_001038672.1 | 4 - 5 | 834 | 79 | |
XM_005158734.4 | XP_005158791.1 | 4 - 5 | 855 | 79 | ||
XM_021467217.1 | XP_021322892.1 | 4 - 5 | 855 | 79 | ||
XM_021467218.1 | XP_021322893.1 | 4 - 5 | 855 | 79 | ||
XM_021467219.1 | XP_021322894.1 | 4 - 5 | 855 | 79 | ||
GenBank mRNA | BC135060.1 | - | 4 - 5 | 833 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |