Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emilin1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045342.1 | NP_001038807.1 | 3 - 4 | 834 | 101 | |
GenBank mRNA | BC121198.1 | - | 3 - 4 | 834 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |