Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045485.2 | NP_001038950.1 | 1 - 2 | 491 | 78 | |
GenBank mRNA | AY929291.1 | - | 1 - 2 | 328 | 78 | |
BC129195.1 | - | 1 - 2 | 491 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |